Merck
  • On the photostability of some blue-emitting derivatives of 2-aminoterephthalic acid and their copolymers with methyl methacrylate
On the photostability of some blue-emitting derivatives of 2-aminoterephthalic acid and their copolymers with methyl methacrylate
Polymer Degradation and Stability (2013)
Miladinova P, et al.