Merck
  • Optimization and validation of an HPLC-photodiode array detector method for determination of organic acids in vinegar
Optimization and validation of an HPLC-photodiode array detector method for determination of organic acids in vinegar
Journal of AOAC (Association of Official Analytical Chemists) International (2015)
Zong Y, et al.