Merck
  • Evolution of tryptophan and its foremost metabolites? concentrations in milk and fermented dairy products
Evolution of tryptophan and its foremost metabolites? concentrations in milk and fermented dairy products
PharmaNutrition (2016)
Bertazzo A, et al.