Merck
  • Can Indirect Herbicide Exposure Modify the Response of the Colorado Potato Beetle to an Organophosphate Insecticide?
Can Indirect Herbicide Exposure Modify the Response of the Colorado Potato Beetle to an Organophosphate Insecticide?
Journal of economic entomology (2019-05-14)
Aigi Margus, Miia Rainio, Leena Lindström
ABSTRACT
Organisms live in complex multivariate environments. In agroecosystems, this complexity is often human-induced as pest individuals can be exposed to many xenobiotics simultaneously. Predicting the effects of multiple stressors can be problematic, as two or more stressors can have interactive effects. Our objective was to investigate whether indirect glyphosate-based herbicide (GBH) exposure of the host plant has interactive effects in combination with an insecticide (azinphos-methyl) on an invasive pest Colorado potato beetle (Leptinotarsa decemlineata Say). We tested the effects of GBH and insecticide on the survival, insecticide target genes expression (acetylcholinesterase genes) and oxidative status biomarkers (glutathione S-transferase [GST], glucose-6-phosphate dehydrogenase [G6PDH], glutathione reductase homolog [GR], glutathione peroxidase homolog [GPx], total glutathione [totGSH], glutathione reduced-oxidized [GSH: GSSG], catalase [CAT], superoxide dismutase [SOD], lipid hydroperoxides). We found that exposure to indirect GBH has no single or interactive effects in combination with the insecticide on larval survival. However, prior exposure to GBH inhibits Ldace1 gene expression by 0.55-fold, which is the target site for the organophosphate and carbamate insecticides. This difference disappears when individuals are exposed to both GBH and insecticide, suggesting an antagonistic effect. On the other hand, oxidative status biomarker scores (PCAs of GPx, GR, and CAT) were decreased when exposed to both stressors, indicating a synergistic effect. Overall, we found that indirect GBH exposure can have both antagonistic and synergistic effects in combination with an insecticide, which should be considered when aiming for an ecologically relevant risk assessment of multiple human-induced stressors.
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt hydrate, ≥93% (HPLC)
Sigma-Aldrich
Glutathione-S-Transferase (GST) Assay Kit, sufficient for 500 multiwell tests
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Gly-Gly, ≥99% (titration)
Sigma-Aldrich
SOD Assay Kit, sufficient for 500 tests
Sigma-Aldrich
Catalase Assay Kit, sufficient for ≥100 tests enzymatic, determination of catalase activity in tissues and cells
Sigma-Aldrich
Glutathione Peroxidase Cellular Activity Assay Kit, Sufficient for 100 colorimetric tests
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Cumene hydroperoxide, technical grade, 80%
Sigma-Aldrich
D-Glucose 6-phosphate sodium salt, ≥98% (HPLC)