Merck
  • Germline NPM1 mutations lead to altered rRNA 2'-O-methylation and cause dyskeratosis congenita.
Germline NPM1 mutations lead to altered rRNA 2'-O-methylation and cause dyskeratosis congenita.
Nature genetics (2019-10-02)
Daphna Nachmani, Anne H Bothmer, Silvia Grisendi, Aldo Mele, Dietmar Bothmer, Jonathan D Lee, Emanuele Monteleone, Ke Cheng, Yang Zhang, Assaf C Bester, Alison Guzzetti, Caitlin A Mitchell, Lourdes M Mendez, Olga Pozdnyakova, Paolo Sportoletti, Maria-Paola Martelli, Tom J Vulliamy, Modi Safra, Schraga Schwartz, Lucio Luzzatto, Olivier Bluteau, Jean Soulier, Robert B Darnell, Brunangelo Falini, Inderjeet Dokal, Keisuke Ito, John G Clohessy, Pier Paolo Pandolfi
ABSTRACT
RNA modifications are emerging as key determinants of gene expression. However, compelling genetic demonstrations of their relevance to human disease are lacking. Here, we link ribosomal RNA 2'-O-methylation (2'-O-Me) to the etiology of dyskeratosis congenita. We identify nucleophosmin (NPM1) as an essential regulator of 2'-O-Me on rRNA by directly binding C/D box small nucleolar RNAs, thereby modulating translation. We demonstrate the importance of 2'-O-Me-regulated translation for cellular growth, differentiation and hematopoietic stem cell maintenance, and show that Npm1 inactivation in adult hematopoietic stem cells results in bone marrow failure. We identify NPM1 germline mutations in patients with dyskeratosis congenita presenting with bone marrow failure and demonstrate that they are deficient in small nucleolar RNA binding. Mice harboring a dyskeratosis congenita germline Npm1 mutation recapitulate both hematological and nonhematological features of dyskeratosis congenita. Thus, our findings indicate that impaired 2'-O-Me can be etiological to human disease.
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
21-Hydroxyprogesterone, ≥97% (HPLC)
Sigma-Aldrich
Denhardt′s Solution 50x, for Northern and Southern blotting, solution
Sigma-Aldrich
Hemin, BioXtra, from Porcine, ≥96.0% (HPLC)
Sigma-Aldrich
5-Bromo-2′-deoxyuridine 5′-triphosphate sodium salt, ≥90%
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, ≥99% (HPLC)
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose and sodium bicarbonate, without L-methionine, L-cystine and L-glutamine, liquid, sterile-filtered, suitable for cell culture