Merck
  • Neutralization of the Positive Charges on Histone Tails by RNA Promotes an Open Chromatin Structure.
Neutralization of the Positive Charges on Histone Tails by RNA Promotes an Open Chromatin Structure.
Cell chemical biology (2019-08-27)
Rositsa Dueva, Karen Akopyan, Chiara Pederiva, Davide Trevisan, Soniya Dhanjal, Arne Lindqvist, Marianne Farnebo
ABSTRACT
RNA associates extensively with chromatin and can influence its structure; however, the potential role of the negative charges of RNA on chromatin structure remains unknown. Here, we demonstrate that RNA prevents precipitation of histones and can attenuate electrostatic interactions between histones and DNA, thereby loosening up the chromatin structure. This effect is independent of the sequence of RNA but dependent on its single-stranded nature, length, concentration, and negative charge. Opening and closure of chromatin by RNA occurs rapidly (within minutes) and passively (in permeabilized cells), in agreement with electrostatics. Accordingly, chromatin compaction following removal of RNA can be prevented by high ionic strength or neutralization of the positively charged histone tails by hyperacetylation. Finally, LINE1 repeat RNAs bind histone H2B and can decondense chromatin. We propose that RNA regulates chromatin opening and closure by neutralizing the positively charged tails of histones, reducing their electrostatic interactions with DNA.
Sigma-Aldrich
Trichostatin A, Ready Made Solution, 5 mM in DMSO, from Streptomyces sp.
Sigma-Aldrich
1-Bromo-3-chloropropane, 99%
Sigma-Aldrich
1-Bromo-3-chloropropane, for isolation of RNA
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Proteinase K from Tritirachium album, buffered aqueous glycerol solution, Molecular Biology, ≥800 units/mL
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Anti-Histone H2B Antibody, clone 5HH2-2A8, ascites fluid, clone 5HH2-2A8, from mouse
Sigma-Aldrich
α-Amanitin, from Amanita phalloides, ≥85% (HPLC), powder