Merck
  • Compositional changes to the ileal microbiome precede the onset of spontaneous ileitis in SHIP deficient mice.
Compositional changes to the ileal microbiome precede the onset of spontaneous ileitis in SHIP deficient mice.
Gut microbes (2019-02-15)
Peter Allan Dobranowski, Chris Tang, Jean Philippe Sauvé, Susan Christine Menzies, Laura May Sly
ABSTRACT
Inflammatory bowel disease, encompassing both ulcerative colitis and Crohn's disease, is characterized by chronic, relapsing-remitting gastrointestinal inflammation of unknown etiology. SHIP deficient mice develop fully penetrant, spontaneous ileitis at 6 weeks of age, and thus offer a tractable model of Crohn's disease-like inflammation. Since disruptions to the microbiome are implicated in the pathogenesis of Crohn's disease, we conducted a 16S rRNA gene survey of the ileum, cecum, colon, and stool contents of SHIP+/+ and SHIP-/- mice. We predicted that diversity and compositional changes would occur after, and possibly prior to, the onset of overt disease. No differences were found in alpha diversity, but significant changes in beta diversity and specific commensal populations were observed in the ileal compartment of SHIP deficient mice after the onset of overt disease. Specifically, reductions in the Bacteroidales taxa, Muribaculum intestinale, and an expansion in Lactobacillus were most notable. In contrast, expansions to bacterial taxa previously associated with inflammation, including Bacteroides, Parabacteroides, and Prevotella were observed in the ilea of SHIP deficient mice prior to the onset of overt disease. Finally, antibiotic treatment reduced the development of intestinal inflammation in SHIP-/- mice. Thus, our findings indicate that SHIP is involved in maintaining ileal microbial homeostasis. These results have broader implications for humans, since reduced SHIP protein levels have been reported in people with Crohn's disease.
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Supelco
Metronidazole, analytical standard
Sigma-Aldrich
Carmine, powder, ≥42.0% (UV-vis)
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP