Merck
  • Phosphatidylserine transport by Ups2-Mdm35 in respiration-active mitochondria.
Phosphatidylserine transport by Ups2-Mdm35 in respiration-active mitochondria.
The Journal of cell biology (2016-06-30)
Non Miyata, Yasunori Watanabe, Yasushi Tamura, Toshiya Endo, Osamu Kuge
ABSTRACT
Phosphatidylethanolamine (PE) is an essential phospholipid for mitochondrial functions and is synthesized mainly by phosphatidylserine (PS) decarboxylase at the mitochondrial inner membrane. In Saccharomyces cerevisiae, PS is synthesized in the endoplasmic reticulum (ER), such that mitochondrial PE synthesis requires PS transport from the ER to the mitochondrial inner membrane. Here, we provide evidence that Ups2-Mdm35, a protein complex localized at the mitochondrial intermembrane space, mediates PS transport for PE synthesis in respiration-active mitochondria. UPS2- and MDM35-null mutations greatly attenuated conversion of PS to PE in yeast cells growing logarithmically under nonfermentable conditions, but not fermentable conditions. A recombinant Ups2-Mdm35 fusion protein exhibited phospholipid-transfer activity between liposomes in vitro. Furthermore, UPS2 expression was elevated under nonfermentable conditions and at the diauxic shift, the metabolic transition from glycolysis to oxidative phosphorylation. These results demonstrate that Ups2-Mdm35 functions as a PS transfer protein and enhances mitochondrial PE synthesis in response to the cellular metabolic state.
Product Description
Avanti
18:1 PA, Avanti Research - A Croda Brand
Avanti
18:1 PS (DOPS), Avanti Research - A Croda Brand
Avanti
18:1 Liss Rhod PE, Avanti Research - A Croda Brand
Avanti
18:1-12:0 NBD PA, Avanti Research - A Croda Brand
Avanti
18:1-12:0 NBD PE, Avanti Research - A Croda Brand 810156C
Avanti
18:1-12:0 NBD PS, Avanti Research - A Croda Brand 810195C
Avanti
18:1-12:0 NBD PC, Avanti Research - A Croda Brand 810133C
Avanti
18:1 Liss Rhod PE, Avanti Research - A Croda Brand
Avanti
18:1-12:0 NBD PC, Avanti Research - A Croda Brand 810133P, powder
Avanti
18:1 PS (DOPS), Avanti Research - A Croda Brand
Avanti
18:1 PA, Avanti Research - A Croda Brand
Avanti
18:1-12:0 NBD PS, Avanti Research - A Croda Brand 810195P, powder
Avanti
18:1-12:0 NBD PE, Avanti Research - A Croda Brand 810156P, powder
Avanti
18:1-12:0 NBD PA, Avanti Research - A Croda Brand 810176C
Avanti
Egg Liss Rhod PE, Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PE (DOPE), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PE (DOPE), Avanti Research - A Croda Brand
Avanti
Egg Liss Rhod PE, Avanti Research - A Croda Brand, powder