Merck
  • Atomic resolution map of the soluble amyloid beta assembly toxic surfaces.
Atomic resolution map of the soluble amyloid beta assembly toxic surfaces.
Chemical science (2019-07-31)
Rashik Ahmed, Michael Akcan, Adree Khondker, Maikel C Rheinstädter, José C Bozelli, Richard M Epand, Vincent Huynh, Ryan G Wylie, Stephen Boulton, Jinfeng Huang, Chris P Verschoor, Giuseppe Melacini
ABSTRACT
Soluble amyloid beta assemblies (Aβ n ) are neurotoxic and play a central role in the early phases of the pathogenesis cascade leading to Alzheimer's disease. However, the current knowledge about the molecular determinants of Aβ n toxicity is at best scant. Here, we comparatively analyze Aβ n prepared in the absence or presence of a catechin library that modulates cellular toxicity. By combining solution NMR with dynamic light scattering, fluorescence spectroscopy, electron microscopy, wide-angle X-ray diffraction and cell viability assays, we identify a cluster of unique molecular signatures that distinguish toxic vs. nontoxic Aβ assemblies. These include the exposure of a hydrophobic surface spanning residues 17-28 and the concurrent shielding of the highly charged N-terminus. We show that the combination of these two dichotomous structural transitions promotes the colocalization and insertion of β-sheet rich Aβ n into the membrane, compromising membrane integrity. These previously elusive toxic surfaces mapped here provide an unprecedented foundation to establish structure-toxicity relationships of Aβ assemblies.
Product Description
Avanti
18:1 (Δ9-Cis) PE (DOPE), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PE (DOPE), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:1 PS (DOPS), Avanti Research - A Croda Brand
Avanti
18:1 PS (DOPS), Avanti Research - A Croda Brand