Merck
  • Variation of the detergent-binding capacity and phospholipid content of membrane proteins when purified in different detergents.
Variation of the detergent-binding capacity and phospholipid content of membrane proteins when purified in different detergents.
Biophysical journal (2014-04-18)
Hüseyin Ilgü, Jean-Marc Jeckelmann, María Salomé Gachet, Rajendra Boggavarapu, Zöhre Ucurum, Jürg Gertsch, Dimitrios Fotiadis
ABSTRACT
Purified membrane proteins are ternary complexes consisting of protein, lipid, and detergent. Information about the amounts of detergent and endogenous phospholipid molecules bound to purified membrane proteins is largely lacking. In this systematic study, three model membrane proteins of different oligomeric states were purified in nine different detergents at commonly used concentrations and characterized biochemically and biophysically. Detergent-binding capacities and phospholipid contents of the model proteins were determined and compared. The insights on ternary complexes obtained from the experimental results, when put into a general context, are summarized as follows. 1), The amount of detergent and 2) the amount of endogenous phospholipids bound to purified membrane proteins are dependent on the size of the hydrophobic lipid-accessible protein surface areas and the physicochemical properties of the detergents used. 3), The size of the detergent and lipid belt surrounding the hydrophobic lipid-accessible surface of purified membrane proteins can be tuned by the appropriate choice of detergent. 4), The detergents n-nonyl-β-D-glucopyranoside and Cymal-5 have exceptional delipidating effects on ternary complexes. 5), The types of endogenous phospholipids bound to membrane proteins can vary depending on the detergent used for solubilization and purification. 6), Furthermore, we demonstrate that size-exclusion chromatography can be a suitable method for estimating the molecular mass of ternary complexes. The findings presented suggest a strategy to control and tune the numbers of detergent and endogenous phospholipid molecules bound to membrane proteins. These two parameters are potentially important for the successul crystallization of membrane proteins for structure determination by crystallographic approaches.
Product Description
Avanti
E. coli Extract Total, Avanti Research - A Croda Brand
Avanti
E. coli Extract Polar, Avanti Research - A Croda Brand
Avanti
E. coli Extract Polar, Avanti Research - A Croda Brand
Avanti
E. coli Extract Total, Avanti Research - A Croda Brand