Merck
How synaptotagmin promotes membrane fusion.
Science (New York, N.Y.) (2007-05-05)
Sascha Martens, Michael M Kozlov, Harvey T McMahon
ABSTRACT
Synaptic vesicles loaded with neurotransmitters are exocytosed in a soluble N-ethylmaleimide-sensitive factor attachment protein receptor (SNARE)-dependent manner after presynaptic depolarization induces calcium ion (Ca2+) influx. The Ca2+ sensor required for fast fusion is synaptotagmin-1. The activation energy of bilayer-bilayer fusion is very high (approximately 40 k(B)T). We found that, in response to Ca2+ binding, synaptotagmin-1 could promote SNARE-mediated fusion by lowering this activation barrier by inducing high positive curvature in target membranes on C2-domain membrane insertion. Thus, synaptotagmin-1 triggers the fusion of docked vesicles by local Ca2+-dependent buckling of the plasma membrane together with the zippering of SNAREs. This mechanism may be widely used in membrane fusion.
Product Description
Avanti
Brain PI(4,5)P2, Avanti Research - A Croda Brand
Avanti
Brain PI(4,5)P2, Avanti Research - A Croda Brand
Avanti
Brain PC, Avanti Research - A Croda Brand 840053P, powder
Avanti
Brain PC, Avanti Research - A Croda Brand 840053C
Avanti
Brain PS, Avanti Research - A Croda Brand
Avanti
Brain PS, Avanti Research - A Croda Brand