Merck
  • Isolation of phosphatidylethanolamine as a solitary cofactor for prion formation in the absence of nucleic acids.
Isolation of phosphatidylethanolamine as a solitary cofactor for prion formation in the absence of nucleic acids.
Proceedings of the National Academy of Sciences of the United States of America (2012-05-16)
Nathan R Deleault, Justin R Piro, Daniel J Walsh, Fei Wang, Jiyan Ma, James C Geoghegan, Surachai Supattapone
ABSTRACT
Infectious prions containing the pathogenic conformer of the mammalian prion protein (PrP(Sc)) can be produced de novo from a mixture of the normal conformer (PrP(C)) with RNA and lipid molecules. Recent reconstitution studies indicate that nucleic acids are not required for the propagation of mouse prions in vitro, suggesting the existence of an alternative prion propagation cofactor in brain tissue. However, the identity and functional properties of this unique cofactor are unknown. Here, we show by purification and reconstitution that the molecule responsible for the nuclease-resistant cofactor activity in brain is endogenous phosphatidylethanolamine (PE). Synthetic PE alone facilitates conversion of purified recombinant (rec)PrP substrate into infectious recPrP(Sc) molecules. Other phospholipids, including phosphatidylcholine, phosphatidylserine, phosphatidylinositol, and phosphatidylglycerol, were unable to facilitate recPrP(Sc) formation in the absence of RNA. PE facilitated the propagation of PrP(Sc) molecules derived from all four different animal species tested including mouse, suggesting that unlike RNA, PE is a promiscuous cofactor for PrP(Sc) formation in vitro. Phospholipase treatment abolished the ability of brain homogenate to reconstitute the propagation of both mouse and hamster PrP(Sc) molecules. Our results identify a single endogenous cofactor able to facilitate the formation of prions from multiple species in the absence of nucleic acids or other polyanions.
Product Description
Avanti
16:0 Lyso PC, 1-palmitoyl-2-hydroxy-sn-glycero-3-phosphocholine, chloroform
Avanti
16:0 Lyso PC, Avanti Research - A Croda Brand
Avanti
Brain PE, Avanti Research - A Croda Brand 840022C
Avanti
Brain PE, Avanti Research - A Croda Brand 840022P, powder
Avanti
Liver PI, Avanti Research - A Croda Brand
Avanti
Liver PI, Avanti Research - A Croda Brand
Avanti
C18(Plasm)-18:1 PE, Avanti Research - A Croda Brand
Avanti
Brain PC, Avanti Research - A Croda Brand 840053C
Avanti
Egg PG, Avanti Research - A Croda Brand
Avanti
Brain PC, Avanti Research - A Croda Brand 840053P, powder
Avanti
Brain PS, Avanti Research - A Croda Brand
Avanti
Brain PS, Avanti Research - A Croda Brand
Avanti
Egg PG, Avanti Research - A Croda Brand
Avanti
16:0 Lyso PE, Avanti Research - A Croda Brand, powder
Avanti
C18(Plasm)-18:1 PE, 1-(1Z-octadecenyl)-2-oleoyl-sn-glycero-3-phosphoethanolamine, powder