Merck
  • Xenopus extract approaches to studying microtubule organization and signaling in cytokinesis.
Xenopus extract approaches to studying microtubule organization and signaling in cytokinesis.
Methods in cell biology (2017-01-10)
C M Field, J F Pelletier, T J Mitchison
ABSTRACT
We report optimized methods for preparing actin-intact Xenopus egg extract. This extract is minimally perturbed, undiluted egg cytoplasm where the cell cycle can be experimentally controlled. It contains abundant organelles and glycogen and supports active metabolism and cytoskeletal dynamics that closely mimic egg physiology. The concentration of the most abundant ∼11,000 proteins is known from mass spectrometry. Actin-intact egg extract can be used for analysis of actin dynamics and interaction of actin with other cytoplasmic systems, as well as microtubule organization. It can be spread as thin layers and naturally depletes oxygen though mitochondrial metabolism, which makes it ideal for fluorescence imaging. When combined with artificial lipid bilayers, it allows reconstitution and analysis of the spatially controlled signaling that positions the cleavage furrow during early cytokinesis. Actin-intact extract is generally useful for probing the biochemistry and biophysics of the large Xenopus egg. Protocols are provided for preparation of actin-intact egg extract, control of the cell cycle, fluorescent probes for cytoskeleton and cytoskeleton-dependent signaling, preparation of glass surfaces for imaging experiments, and immunodepletion to probe the role of specific proteins and protein complexes. We also describe methods for adding supported lipid bilayers to mimic the plasma membrane and for confining in microfluidic droplets to explore size scaling issues.
Sigma-Aldrich
Anti-phospho-Ser/Thr-Pro MPM-2 Antibody, clone MPM-2, Upstate®, from mouse
Avanti
Brain PI(4,5)P2, Avanti Research - A Croda Brand
Avanti
Brain PI(4,5)P2, Avanti Research - A Croda Brand
Sigma-Aldrich
Leupeptin, microbial, ≥90% (HPLC)
Sigma-Aldrich
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
Sigma-Aldrich
Pepstatin A, microbial, ≥90% (HPLC)
Sigma-Aldrich
bisBenzimide H 33258, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC)
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-α-Lysophosphatidylcholine from egg yolk, ≥99%, Type I, powder
Sigma-Aldrich
Spermidine trihydrochloride, ≥98% (TLC)
Avanti
Brain PC, Avanti Research - A Croda Brand 840053P, powder
Avanti
Liver PI, Avanti Research - A Croda Brand
Avanti
Brain PC, Avanti Research - A Croda Brand 840053C
Avanti
16:0 Liss Rhod PE, Avanti Research - A Croda Brand
Avanti
Brain PS, Avanti Research - A Croda Brand
Avanti
Brain PS, Avanti Research - A Croda Brand
Avanti
Liver PI, Avanti Research - A Croda Brand
Avanti
16:0 Liss Rhod PE, Avanti Research - A Croda Brand