Merck
  • Human Papillomavirus Oncogene Manipulation Using Clustered Regularly Interspersed Short Palindromic Repeats/Cas9 Delivered by pH-Sensitive Cationic Liposomes.
Human Papillomavirus Oncogene Manipulation Using Clustered Regularly Interspersed Short Palindromic Repeats/Cas9 Delivered by pH-Sensitive Cationic Liposomes.
Human gene therapy (2020-01-25)
Shuai Zhen, Yan Liu, Jiaojiao Lu, Xiaoqian Tuo, Xiling Yang, Hong Chen, Wei Chen, Xu Li
ABSTRACT
Clustered regularly interspaced short palindromic repeat (CRISPR)/CRISPR-associated protein 9 (Cas9) technology enables targeted gene editing, but cancer gene therapy with this approach requires improvements to enable safe and efficient delivery of CRISPR/Cas9 to tumors. We developed and evaluated a self-assembled liposome to selectively deliver CRISPR/Cas9 to cancer tissues. Our CRISPR/Cas9 system effectively inhibited proliferation of human papillomavirus (HPV) 16-positive cervical cancer cells and induced apoptosis by inactivating the HR-HPV16E6/E7 oncogene. Based on this system, we prepared a long-circulating pH-sensitive cationic nano-liposome complex with a high cell targeting and gene knockout rate. Intratumoral injection of cationic liposomes targeted to splicing HPV16 E6/E7 in nude mice significantly inhibited tumor growth without significant toxicity in vivo. Liposomes that targeted HPV16 E6/E7 splicing were established as a basis for treatment of HPV16-positive cervical cancer drug candidates. Our study demonstrates that this liposome offers an efficient delivery system for nonviral gene editing, adding to the armamentarium of gene editing tools to advance safe and effective precision medicine-based cancer therapeutics.
Product Description
Avanti
18:1 (Δ9-Cis) PE (DOPE), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PE (DOPE), Avanti Research - A Croda Brand
Avanti
18:1 TAP (DOTAP), Avanti Research - A Croda Brand
Avanti
18:1 TAP (DOTAP), Avanti Research - A Croda Brand
Avanti
18:0 PEG2000 PE, Avanti Research - A Croda Brand 880120C
Avanti
18:0 PEG2000 PE (Ammonium Salt), Avanti Research - A Croda Brand