Merck
  • Biased Signaling of the G-Protein-Coupled Receptor β2AR Is Governed by Conformational Exchange Kinetics.
Biased Signaling of the G-Protein-Coupled Receptor β2AR Is Governed by Conformational Exchange Kinetics.
Structure (London, England : 1993) (2020-01-25)
Rajan Lamichhane, Jeffrey J Liu, Kate L White, Vsevolod Katritch, Raymond C Stevens, Kurt Wüthrich, David P Millar
ABSTRACT
G-protein-coupled receptors (GPCRs) mediate a wide range of human physiological functions by transducing extracellular ligand binding events into intracellular responses. GPCRs can activate parallel, independent signaling pathways mediated by G proteins or β-arrestins. Whereas "balanced" agonists activate both pathways equally, "biased" agonists dominantly activate one pathway, which is of interest for designing GPCR-targeting drugs because it may mitigate undesirable side effects. Previous studies demonstrated that β-arrestin activation is associated with transmembrane helix VII (TM VII) of GPCRs. Here, single-molecule fluorescence spectroscopy with the β2-adrenergic receptor (β2AR) in the ligand-free state showed that TM VII spontaneously fluctuates between one inactive and one active-like conformation. The presence of the β-arrestin-biased agonist isoetharine prolongs the dwell time of TM VII in the active-like conformation compared with the balanced agonist formoterol, suggesting that ligands can induce signaling bias by modulating the kinetics of receptor conformational exchange.
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type VII, lyophilized powder, ≥100,000 units/g solid (without added oxygen)
Sigma-Aldrich
Cholesteryl hemisuccinate tris salt, anionic detergent
Avanti
16:0-18:1 PS (POPS), Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PS (POPS), Avanti Research - A Croda Brand
Sigma-Aldrich
Maleimide, 99%
Avanti
18:1 Biotinyl Cap PE, Avanti Research - A Croda Brand 870273P, powder
Avanti
18:1 Biotinyl Cap PE, Avanti Research - A Croda Brand 870273C