Merck
  • Selective Persulfide Detection Reveals Evolutionarily Conserved Antiaging Effects of S-Sulfhydration.
Selective Persulfide Detection Reveals Evolutionarily Conserved Antiaging Effects of S-Sulfhydration.
Cell metabolism (2019-11-19)
Jasmina Zivanovic, Emilia Kouroussis, Joshua B Kohl, Bikash Adhikari, Biljana Bursac, Sonia Schott-Roux, Dunja Petrovic, Jan Lj Miljkovic, Daniel Thomas-Lopez, Youngeun Jung, Marko Miler, Sarah Mitchell, Verica Milosevic, Jose Eduardo Gomes, Moran Benhar, Bruno Gonzalez-Zorn, Ivana Ivanovic-Burmazovic, Roberta Torregrossa, James R Mitchell, Matthew Whiteman, Guenter Schwarz, Solomon H Snyder, Bindu D Paul, Kate S Carroll, Milos R Filipovic
ABSTRACT
Life on Earth emerged in a hydrogen sulfide (H2S)-rich environment eons ago and with it protein persulfidation mediated by H2S evolved as a signaling mechanism. Protein persulfidation (S-sulfhydration) is a post-translational modification of reactive cysteine residues, which modulate protein structure and/or function. Persulfides are difficult to label and study due to their reactivity and similarity with cysteine. Here, we report a facile strategy for chemoselective persulfide bioconjugation using dimedone-based probes, to achieve highly selective, rapid, and robust persulfide labeling in biological samples with broad utility. Using this method, we show persulfidation is an evolutionarily conserved modification and waves of persulfidation are employed by cells to resolve sulfenylation and prevent irreversible cysteine overoxidation preserving protein function. We report an age-associated decline in persulfidation that is conserved across evolutionary boundaries. Accordingly, dietary or pharmacological interventions to increase persulfidation associate with increased longevity and improved capacity to cope with stress stimuli.
Sigma-Aldrich
Anti-MPST antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
O-(Carboxymethyl)hydroxylamine hemihydrochloride, 98%
Sigma-Aldrich
Sodium sulfide hydrate, ≥60%, scales
Sigma-Aldrich
Trypsin from porcine pancreas, Proteomics Grade, BioReagent, Dimethylated
Sigma-Aldrich
Sodium sulfide
Sigma-Aldrich
5,5-Dimethyl-1,3-cyclohexanedione, 95%
Sigma-Aldrich
Insulin human, meets USP testing specifications
Supelco
Paraquat dichloride hydrate, PESTANAL®, analytical standard
Sigma-Aldrich
Chelex® 100 sodium form, 50-100 mesh (dry)
Sigma-Aldrich
Monensin sodium salt, 90-95% (TLC)
Sigma-Aldrich
DL-Propargylglycine, cystathionine γ-lyase inhibitor
Sigma-Aldrich
Albumin from human serum, lyophilized powder, essentially fatty acid free
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Glyceraldehyde-3-phosphate Dehydrogenase from rabbit muscle, lyophilized powder, ≥75 units/mg protein
Sigma-Aldrich
L-Cysteine S-sulfate, ≥98% (TLC)
Sigma-Aldrich
Monoclonal Anti-Biotin−Peroxidase antibody produced in mouse, clone BN-34, purified immunoglobulin
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
L-Ascorbic acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Erastin, ≥98% (HPLC)
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-SQRDL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
5-Fluoro-2′-deoxyuridine, thymidylate synthase inhibitor
Sigma-Aldrich
Sodium (meta)arsenite, ≥90%
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, clone D66, purified from hybridoma cell culture
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Thioredoxin Reductase from rat liver, buffered aqueous glycerol solution, ≥100 units/mg protein (Bradford)
Sigma-Aldrich
4-Chloro-7-nitrobenzofurazan, 98%