Merck
  • A novel transporter of SLC22 family specifically transports prostaglandins and co-localizes with 15-hydroxyprostaglandin dehydrogenase in renal proximal tubules.
A novel transporter of SLC22 family specifically transports prostaglandins and co-localizes with 15-hydroxyprostaglandin dehydrogenase in renal proximal tubules.
The Journal of biological chemistry (2010-05-08)
Katsuko Shiraya, Taku Hirata, Ryo Hatano, Shushi Nagamori, Pattama Wiriyasermkul, Promsuk Jutabha, Mitsunobu Matsubara, Shigeaki Muto, Hidekazu Tanaka, Shinji Asano, Naohiko Anzai, Hitoshi Endou, Akira Yamada, Hiroyuki Sakurai, Yoshikatsu Kanai
ABSTRACT
We identified a novel prostaglandin (PG)-specific organic anion transporter (OAT) in the OAT group of the SLC22 family. The transporter designated OAT-PG from mouse kidney exhibited Na(+)-independent and saturable transport of PGE(2) when expressed in a proximal tubule cell line (S(2)). Unusual for OAT members, OAT-PG showed narrow substrate selectivity and high affinity for a specific subset of PGs, including PGE(2), PGF(2alpha), and PGD(2). Similar to PGE(2) receptor and PGT, a structurally distinct PG transporter, OAT-PG requires for its substrates an alpha-carboxyl group, with a double bond between C13 and C14 as well as a (S)-hydroxyl group at C15. Unlike the PGE(2) receptor, however, the hydroxyl group at C11 in a cyclopentane ring is not essential for OAT-PG substrates. Addition of a hydroxyl group at C19 or C20 impairs the interaction with OAT-PG, whereas an ethyl group at C20 enhances the interaction, suggesting the importance of hydrophobicity around the omega-tail tip forming a "hydrophobic core" accompanied by a negative charge, which is essential for substrates of OAT members. OAT-PG-mediated transport is concentrative in nature, although OAT-PG mediates both facilitative and exchange transport. OAT-PG is kidney-specific and localized on the basolateral membrane of proximal tubules where a PG-inactivating enzyme, 15-hydroxyprostaglandin dehydrogenase, is expressed. Because of the fact that 15-keto-PGE(2), the metabolite of PGE(2) produced by 15-hydroxyprostaglandin dehydrogenase, is not a substrate of OAT-PG, the transport-metabolism coupling would make unidirectional PGE(2) transport more efficient. By removing extracellular PGE(2), OAT-PG is proposed to be involved in the local PGE(2) clearance and metabolism for the inactivation of PG signals in the kidney cortex.
Sigma-Aldrich
15-Deoxy-Δ12,14-prostaglandin J2, ≥95% (HPLC), 1 mg/mL in methyl acetate
Sigma-Aldrich
Prostaglandin I2 sodium salt, ≥96% (HPLC), synthetic, powder
Sigma-Aldrich
Prostaglandin E1, ≥98% (HPLC), synthetic
Sigma-Aldrich
Prostaglandin F tris salt, synthetic, suitable for cell culture
Sigma-Aldrich
Prostaglandin D2, ≥95%, synthetic
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic