- Cyanobacterial microcystin-LR is a potent and specific inhibitor of protein phosphatases 1 and 2A from both mammals and higher plants.
Cyanobacterial microcystin-LR is a potent and specific inhibitor of protein phosphatases 1 and 2A from both mammals and higher plants.
FEBS letters (1990-05-21)
C MacKintosh, K A Beattie, S Klumpp, P Cohen, G A Codd
PMID2162782
ABSTRACT
The cyclic heptapeptide, microcystin-LR, inhibits protein phosphatases 1 (PP1) and 2A (PP2A) with Ki values below 0.1 nM. Protein phosphatase 2B is inhibited 1000-fold less potently, while six other phosphatases and eight protein kinases tested are unaffected. These results are strikingly similar to those obtained with the tumour promoter okadaic acid. We establish that okadaic acid prevents the binding of microcystin-LR to PP2A, and that protein inhibitors 1 and 2 prevent the binding of microcystin-LR to PP1. We discuss the possibility that inhibition of PP1 and PP2A accounts for the extreme toxicity of microcystin-LR, and indicate its potential value in the detection and analysis of protein kinases and phosphatases.