Merck
  • Cryo-Electron Tomography and Proteomics studies of centrosomes from differentiated quiescent thymocytes.
Cryo-Electron Tomography and Proteomics studies of centrosomes from differentiated quiescent thymocytes.
Scientific reports (2019-05-12)
Johan Busselez, Francisco Javier Chichón, Maria Josefa Rodríguez, Adan Alpízar, Séverine Isabelle Gharbi, Mònica Franch, Roberto Melero, Alberto Paradela, José L Carrascosa, José-Maria Carazo
ABSTRACT
We have used cryo Electron Tomography, proteomics and immunolabeling to study centrosomes isolated from the young lamb thymus, an efficient source of quiescent differentiated cells. We compared the proteome of thymocyte centrosomes to data published for KE37 cells, focusing on proteins associated with centriole disengagement and centrosome separation. The data obtained enhances our understanding of the protein system joining the centrioles, a system comprised of a branched network of fibers linked to an apparently amorphous density that was partially characterized here. A number of proteins were localized to the amorphous density by immunolabeling (C-NAP1, cohesin SMC1, condensin SMC4 and NCAPD2), yet not DNA. In conjuction, these data not only extend our understanding of centrosomes but they will help refine the model that focus on the protein system associated with the centriolar junction.
Sigma-Aldrich
Monoclonal Anti-CNAP1 antibody produced in mouse, clone 4C9, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid
Sigma-Aldrich
Anti-DNA Antibody, clone AC-30-10, clone AC-30-10, Chemicon®, from mouse