Merck
  • Disruption of the NF-κB/NLRP3 connection by melatonin requires retinoid-related orphan receptor-α and blocks the septic response in mice.
Disruption of the NF-κB/NLRP3 connection by melatonin requires retinoid-related orphan receptor-α and blocks the septic response in mice.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2015-06-06)
José A García, Huayqui Volt, Carmen Venegas, Carolina Doerrier, Germaine Escames, Luis C López, Darío Acuña-Castroviejo
ABSTRACT
We determined the NF-κB- and NOD-like receptor (NLR)P3-dependent molecular mechanisms involved in sepsis and evaluated the role of retinoid-related orphan receptor (ROR)-α in melatonin's anti-inflammatory actions. Western blot, RT-PCR, ELISA, and spectrophotometric analysis revealed that NF-κB and NLRP3 closely interact, leading to proinflammatory and pro-oxidant status in heart tissue of septic C57BL/6J mice. Moreover, mitochondrial oxygen consumption was reduced by 80% in septic mice. In vivo and in vitro analysis showed that melatonin administration blunts NF-κB transcriptional activity through a sirtuin1-dependent NF-κB deacetylation in septic mice. Melatonin also decreased NF-κB-dependent proinflammatory response and restored redox balance and mitochondrial homeostasis, thus inhibiting the NLRP3 inflammasome. In an important finding, the inhibition of NF-κB by melatonin, but not that of NLRP3, was blunted in RORα (sg/sg) mice, indicating that functional RORα transcription factor is necessary for the initiation of the innate immune response against inflammation. Our results are evidence of the NF-κB/NLRP3 connection during sepsis and identify NLRP3 as a novel molecular target for melatonin. The multiple molecular targets of melatonin in this study explain its potent anti-inflammatory efficacy against systemic innate immune activation and herald a promising therapeutic application for melatonin in the treatment of sepsis.
Sigma-Aldrich
Anti-SIRT1 Antibody, clone 10E4, clone 10-E-4, from mouse
Sigma-Aldrich
Anti-IKKβ Antibody, clone 10AG2, clone 10AG2, Upstate®, from mouse
Sigma-Aldrich
Anti-IKKα Antibody, clone 14A231, clone 14A231, Upstate®, from mouse