Merck
  • Dietary conjugated linoleic acid reduces PGE2 release and interstitial injury in rat polycystic kidney disease.
Dietary conjugated linoleic acid reduces PGE2 release and interstitial injury in rat polycystic kidney disease.
Kidney international (2003-09-13)
Malcolm R Ogborn, Evan Nitschmann, Neda Bankovic-Calic, Hope A Weiler, Shirley Fitzpatrick-Wong, Harold M Aukema
ABSTRACT
Conjugated linoleic acid (CLA) describes positional isomers of linoleic acid (LA). Experimental health benefits of CLA include amelioration of malignancy and inflammatory disease and reduction of adiposity. The Han:SPRD-cy rat model of polycystic kidney disease (PKD) features prominent renal interstitial inflammation and fibrosis that is amenable to dietary modification. We studied CLA supplementation in the modification of inflammatory outcomes in the Han:SPRD-cy rat. Male offspring of Han:SPRD-cy heterozygotes were fed diets, using corn oil or corn oil with a CLA enriched oil (1% of diet by weight as CLA). After 8 weeks, measurements included renal function and morphometry, ex vivo release of renal prostaglandin E2 (PGE2), and renal and hepatic tissue fatty acid profiles. Urine creatinine was significantly higher in PKD animals fed CLA (P = 0.004), but differences in serum creatinine and creatinine clearance did not quite reach significance in PKD animals. CLA feeding reduced interstitial inflammation (P < 0.001), fibrosis (P = 0.03), and renal PGE2 release (P = 0.02). Cystic change and oxidized low-density lipoprotein (LDL) staining did not change significantly. CLA feeding produced increased renal and hepatic CLA isomers. Hepatic, but not renal, LA proportion was reduced on the CLA diet. The renal proportion of the PGE2 precursor, arachidonic acid (AA), was not changed by diet, but hepatic AA proportion increased significantly with CLA feeding (P= 0.009). CLA reduces renal production of PGE2, without reduced availability of the precursor fatty acid, AA. Short-term feeding of CLA to Han:SPRD-cy rats also has significant renal anti-inflammatory and antifibrotic effects. As inflammation and fibrosis are important components of the progression of chronic renal injury, CLA may be a useful agent in dietary amelioration of renal disease.
Sigma-Aldrich
Anti-Macrophages/Monocytes Antibody, clone ED-1, clone ED-1, Chemicon®, from mouse
Sigma-Aldrich
Anti-Low Density Lipoprotein Antibody, copper oxidized, serum, Chemicon®