Merck
  • Appearance and heterochromatin localization of HP1α in early mouse embryos depends on cytoplasmic clock and H3S10 phosphorylation.
Appearance and heterochromatin localization of HP1α in early mouse embryos depends on cytoplasmic clock and H3S10 phosphorylation.
Cell cycle (Georgetown, Tex.) (2012-05-25)
Maciej Meglicki, Marta Teperek-Tkacz, Ewa Borsuk
ABSTRACT
Pericentric constitutive heterochromatin surrounds centromeric regions and is important for centromere function and chromatid cohesion. HP1 (heterochromatin protein 1), a homolog of yeast Swi6, has been shown to be indispensible for proper heterochromatin structure and function. In mammalian somatic cells, two HP1 isoforms, HP1α and HP1β, are constitutively present in pericentric heterochromatin until late G 2, when they dissociate from heterochromatin. Subsequently, they re-associate with heterochromatin at late anaphase. In one-cell mouse embryos, pericentric heterochromatin has a unique configuration and features. It does not form heterochromatin clusters observed in somatic cells and known as chromocenters. Instead, in both pronuclei, it surrounds nucleolar precursor bodies (NBPs), forming ring-like structures. These regions contain HP1β but lack HP1α in both pronuclei. In subsequent interphases, HP1β is constitutively found in heterochromatin until the blastocyst stage. It is not known when HP1α appears and what is its function in early mouse embryos. Here, we show that HP1α appears for the first time at late S phase of two-cell stage, at the time when pericentric heterochromatin is replicated. Its appearance is regulated at the level of translation. In two-cell embryos, the amount of HP1α that can bind to these regions is regulated by phosphorylation of serine 10 of histone H3 (H3S10Ph). Elimination of HP1α by siRNA interfered with centromere relocation from heterochromatin surrounding NPBs to pro-chromocenters at the two-cell stage but did not affect preimplantation develoment to the blastocyst stage.
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
Anti-Heterochromatin Protein-1 α Antibody, clone 2HP-1H5, ascites fluid, clone 2HP-1H5, Chemicon®
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys9) Antibody, Upstate®, from rabbit
Sigma-Aldrich
α-Amanitin, from Amanita phalloides, ≥85% (HPLC), powder
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder