Merck
  • Vitamin E forms inhibit IL-13/STAT6-induced eotaxin-3 secretion by up-regulation of PAR4, an endogenous inhibitor of atypical PKC in human lung epithelial cells.
Vitamin E forms inhibit IL-13/STAT6-induced eotaxin-3 secretion by up-regulation of PAR4, an endogenous inhibitor of atypical PKC in human lung epithelial cells.
The Journal of nutritional biochemistry (2011-07-19)
Yun Wang, Michelle Moreland, James G Wagner, Bruce N Ames, Beate Illek, David B Peden, Qing Jiang
ABSTRACT
Eotaxin-3 (CCL-26), a potent chemokine for eosinophil recruitment and contributing significantly to the pathogenesis of asthma, is secreted by lung epithelial cells in response to T helper 2 cytokines including interleukin 13 (IL-13). Here we showed that vitamin E forms, but not their metabolites, differentially inhibited IL-13-stimulated generation of eotaxin-3 in human lung epithelial A549 cells. The relative inhibitory potency was γ-tocotrienol (γ-TE) (IC50 ~15 μM)>γ-tocopherol, δ-tocopherol (IC50 ~25-50 μM)>α-tocopherol. Consistent with suppression of eotaxin, γ-TE treatment impaired IL-13-induced phosphorylation of STAT6, the key transcription factor for activation of eotaxin expression, and consequently blocked IL-13-stimulated DNA-binding activity of STAT6. In search of the upstream target of γTE by using inhibitor and siRNA approaches, we discovered that the atypical protein kinase C (aPKC) signaling, instead of classical PKC, p38 MAPK, JNK or ERK, played a critical role in IL-13-stimulated eotaxin generation and STAT6 activation. While showing no obvious effect on aPKC expression or phosphorylation, γ-TE treatment resulted in increased expression of prostate-apoptosis-response 4 (PAR4), an endogenous negative regulator of aPKCs. Importantly, γ-TE treatment led to enhanced formation of aPKC/PAR4 complex that is known to reduce aPKC activity via protein-protein crosstalk. Our study demonstrated that γ-TE inhibited IL-13/STAT6-activated eotaxin secretion via up-regulation of PAR4 expression and enhancement of aPKC-PAR4 complex formation. These results support the notion that specific vitamin E forms may be useful anti-asthmatic agents.
Sigma-Aldrich
(+)-δ-Tocopherol, ≥90%
Sigma-Aldrich
(+)-γ-Tocopherol, ≥96% (HPLC)
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
α-Tocopherol, ≥95.5%