Merck
  • A nanotubular metal-organic framework with permanent porosity: structure analysis and gas sorption studies
A nanotubular metal-organic framework with permanent porosity: structure analysis and gas sorption studies
Chemical Communications (Cambridge, England) (2009)
Ma S, et al.