Merck
  • Single-Cell RNA Sequencing Reveals Stromal Evolution into LRRC15+ Myofibroblasts as a Determinant of Patient Response to Cancer Immunotherapy.
Single-Cell RNA Sequencing Reveals Stromal Evolution into LRRC15+ Myofibroblasts as a Determinant of Patient Response to Cancer Immunotherapy.
Cancer discovery (2019-11-09)
Claudia X Dominguez, Sören Müller, Shilpa Keerthivasan, Hartmut Koeppen, Jeffrey Hung, Sarah Gierke, Beatrice Breart, Oded Foreman, Travis W Bainbridge, Alessandra Castiglioni, Yasin Senbabaoglu, Zora Modrusan, Yuxin Liang, Melissa R Junttila, Christiaan Klijn, Richard Bourgon, Shannon J Turley
ABSTRACT
With only a fraction of patients responding to cancer immunotherapy, a better understanding of the entire tumor microenvironment is needed. Using single-cell transcriptomics, we chart the fibroblastic landscape during pancreatic ductal adenocarcinoma (PDAC) progression in animal models. We identify a population of carcinoma-associated fibroblasts (CAF) that are programmed by TGFβ and express the leucine-rich repeat containing 15 (LRRC15) protein. These LRRC15+ CAFs surround tumor islets and are absent from normal pancreatic tissue. The presence of LRRC15+ CAFs in human patients was confirmed in >80,000 single cells from 22 patients with PDAC as well as by using IHC on samples from 70 patients. Furthermore, immunotherapy clinical trials comprising more than 600 patients across six cancer types revealed elevated levels of the LRRC15+ CAF signature correlated with poor response to anti-PD-L1 therapy. This work has important implications for targeting nonimmune elements of the tumor microenvironment to boost responses of patients with cancer to immune checkpoint blockade therapy. SIGNIFICANCE: This study describes the single-cell landscape of CAFs in pancreatic cancer during in vivo tumor evolution. A TGFβ-driven, LRRC15+ CAF lineage is associated with poor outcome in immunotherapy trial data comprising multiple solid-tumor entities and represents a target for combinatorial therapy.This article is highlighted in the In This Issue feature, p. 161.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid
Sigma-Aldrich
Gey′s Balanced Salt Solution, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
OptiPrep Density Gradient Medium, used for cell and subcellular organelle isolation
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Calcein Blue, fluorescent dye
Roche
Trypsin Inhibitor, from soybean
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
Human Serum, from platelet poor human plasma, sterile-filtered, (mycoplasma tested, virus tested)