Merck
  • iHi-C 2.0: A simple approach for mapping native spatial chromatin organisation from low cell numbers.
iHi-C 2.0: A simple approach for mapping native spatial chromatin organisation from low cell numbers.
Methods (San Diego, Calif.) (2019-07-10)
Athanasia Mizi, Eduardo Gade Gusmao, Argyris Papantonis
ABSTRACT
Genome organization is now understood to be tightly linked to all genomic functions. Thus, the high-resolution mapping of higher-order chromosomal structures via 3C-based approaches has become an integral tool for studying transcriptional and cell cycle regulation, signaling effects or disease onset. Nonetheless, 3C-based protocols are not without caveats, like dependencies on fixation conditions, restriction enzyme pervasiveness in crosslinked chromatin and ligation efficiency. To address some of these caveats, we describe here the streamlined iHi-C 2.0 protocol that allows for the genome-wide interrogation of native spatial chromatin contacts without a need for chemical fixation. This approach improves ligation efficiency and presents minimal material losses, and is thus suitable for analysing samples with limiting cell numbers. Following high throughput sequencing, iHi-C 2.0 generates high signal-to-noise and focal maps of the interactions within and between mammalian chromosomes under native conditions.
Sigma-Aldrich
Potassium chloride, Molecular Biology, ≥99.0%
Sigma-Aldrich
Agarose, BioReagent, Molecular Biology, low EEO
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, suitable for (for molecular biology), Non-acetylated, Molecular Biology
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Molecular Biology, ≥70 Kunitz units/mg protein, lyophilized
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 8,000
Sigma-Aldrich
Phenol:Chloroform:Isoamyl Alcohol 25:24:1 Saturated with 10 mM Tris, pH 8.0, 1 mM EDTA, Supplied with Equilibration buffer, Molecular Biology
Sigma-Aldrich
Sodium phosphate dibasic, Molecular Biology, ≥98.5% (titration)
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, Molecular Biology, BioUltra, ≥30 units/mg protein
Sigma-Aldrich
Potassium acetate, Molecular Biology, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, Grade I, ≥99%, from microbial
Sigma-Aldrich
Magnesium chloride hexahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
β-Glycerophosphate disodium salt hydrate, BioUltra, suitable for cell culture, suitable for plant cell culture, ≥99% (titration)