Merck
PT-141: a melanocortin agonist for the treatment of sexual dysfunction.
Annals of the New York Academy of Sciences (2003-07-10)
P B Molinoff, A M Shadiack, D Earle, L E Diamond, C Y Quon
ABSTRACT
PT-141, a synthetic peptide analogue of alpha-MSH, is an agonist at melanocortin receptors including the MC3R and MC4R, which are expressed primarily in the central nervous system. Administration of PT-141 to rats and nonhuman primates results in penile erections. Systemic administration of PT-141 to rats activates neurons in the hypothalamus as shown by an increase in c-Fos immunoreactivity. Neurons in the same region of the central nervous system take up pseudorabies virus injected into the corpus cavernosum of the rat penis. Administration of PT-141 to normal men and to patients with erectile dysfunction resulted in a rapid dose-dependent increase in erectile activity. The results suggest that PT-141 holds promise as a new treatment for sexual dysfunction.
Sigma-Aldrich
Bremelanotide acetate, ≥98% (HPLC)
Pricing and availability is not currently available.