Merck
  • The Ionophoric Activity of a Pro-Apoptotic VEGF165 Fragment on HUVEC Cells.
The Ionophoric Activity of a Pro-Apoptotic VEGF165 Fragment on HUVEC Cells.
International journal of molecular sciences (2020-04-25)
Stefania Zimbone, Anna M Santoro, Diego La Mendola, Chiara Giacomelli, Maria L Trincavelli, Marianna F Tomasello, Danilo Milardi, Sara García-Viñuales, Michele F M Sciacca, Claudia Martini, Giulia Grasso
ABSTRACT
Copper plays an important role as a regulator in many pathologies involving the angiogenesis process. In cancerogenesis, tumor progression, and angiogenic diseases, copper homeostasis is altered. Although many details in the pathways involved are still unknown, some copper-specific ligands have been successfully used as therapeutic agents. Copper-binding peptides able to modulate angiogenesis represent a possible way to value new drugs. We previously reported that a fragment (VEGF73-101) of vascular endothelial growth factor (VEGF165), a potent angiogenic, induced an apoptotic effect on human umbilical vein endothelial cells. The aim of this study was to investigate the putative copper ionophoric activity of VEGF73-101, as well as establish a relationship between the structure of the peptide fragment and the cytotoxic activity in the presence of copper(II) ions. Here, we studied the stoichiometry and the conformation of the VEGF73-101/Cu(II) complexes and some of its mutated peptides by electrospray ionization mass spectrometry and circular dichroism spectroscopy. Furthermore, we evaluated the effect of all peptides in the absence and presence of copper ions by cell viability and cytofuorimetric assays. The obtained results suggest that VEGF73-101 could be considered an interesting candidate in the development of new molecules with ionophoric properties as agents in antiangiogenic therapeutic approaches.
Product Description
Avanti
16:0-18:1 PS (POPS), Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PS (POPS), Avanti Research - A Croda Brand
CellCrown inserts, 6 well plate inserts with 1.0 μm polycarbonate filter, sterile
Avanti
16:0-18:1 PE, Avanti Research - A Croda Brand