Merck
  • Structural elucidation of two types of novel glycosphingolipids in three strains of Skeletonema by liquid chromatography coupled with mass spectrometry.
Structural elucidation of two types of novel glycosphingolipids in three strains of Skeletonema by liquid chromatography coupled with mass spectrometry.
Rapid communications in mass spectrometry : RCM (2013-06-01)
Fang Zhao, Jilin Xu, Juanjuan Chen, Xiaojun Yan, Chengxu Zhou, Shuang Li, Xiaoyan Xu, Fangting Ye
ABSTRACT
Glycosphingolipids (GSLs) are important components of the cell membrane; however, little is known about GSLs in microalgae. We analyzed GSLs in three strains of Skeletonema microalgae by liquid chromatography coupled with mass spectrometry. Total lipid was extracted from Skeletonema microalgae. Separation of lipids was achieved via reversed-phase liquid chromatography, and mass spectrometric analysis of the lithium adduct of GSLs was conducted to determine their structures. Two types of novel glycosphingolipids were identified from three strains of Skeletonema microalgae. The N-acyl groups were primarily long-chain saturated or monoenoic fatty acids (monounsaturated fatty acids) with 16C and 20 to 24C. The sphingoid long-chain bases (LCB) were sphingosine, sphingadienine and sphingatrienine. The saccharide polar head groups of glycosphingolipids were disaccharide (heptose-hexose) or trisaccharide (heptose-hexose-hexose). Semi-quantitative analysis of GLSs in the three strains of microalgae showed that GSL contents ranged from 0.09 to 8.79 nmol/mg dry microalgae powder. A qualitative method was developed for the identification of GSLs in microalgae. Two types of novel GSLs were identified from three strains of Skeletonema, which might have important biological functions. It could also provide a reliable tool for chemotaxonomy of microalgae.
Product Description
Avanti
C24 Lactosyl(β) Ceramide (d18:1/24:0), Avanti Research - A Croda Brand 860577P, powder
Pricing and availability is not currently available.