Merck
  • Gut-seeded α-synuclein fibrils promote gut dysfunction and brain pathology specifically in aged mice.
Gut-seeded α-synuclein fibrils promote gut dysfunction and brain pathology specifically in aged mice.
Nature neuroscience (2020-02-19)
Collin Challis, Acacia Hori, Timothy R Sampson, Bryan B Yoo, Rosemary C Challis, Adam M Hamilton, Sarkis K Mazmanian, Laura A Volpicelli-Daley, Viviana Gradinaru
ABSTRACT
Parkinson's disease is a synucleinopathy that is characterized by motor dysfunction, death of midbrain dopaminergic neurons and accumulation of α-synuclein (α-Syn) aggregates. Evidence suggests that α-Syn aggregation can originate in peripheral tissues and progress to the brain via autonomic fibers. We tested this by inoculating the duodenal wall of mice with α-Syn preformed fibrils. Following inoculation, we observed gastrointestinal deficits and physiological changes to the enteric nervous system. Using the AAV-PHP.S capsid to target the lysosomal enzyme glucocerebrosidase for peripheral gene transfer, we found that α-Syn pathology is reduced due to the increased expression of this protein. Lastly, inoculation of α-Syn fibrils in aged mice, but not younger mice, resulted in progression of α-Syn histopathology to the midbrain and subsequent motor defects. Our results characterize peripheral synucleinopathy in prodromal Parkinson's disease and explore cellular mechanisms for the gut-to-brain progression of α-Syn pathology.
Sigma-Aldrich
5-Ethynyl-2′-deoxyuridine, 95%
Sigma-Aldrich
Fast Green FCF, Dye content ≥85 %