Merck
  • Dual Plasmepsin-Targeting Antimalarial Agents Disrupt Multiple Stages of the Malaria Parasite Life Cycle.
Dual Plasmepsin-Targeting Antimalarial Agents Disrupt Multiple Stages of the Malaria Parasite Life Cycle.
Cell host & microbe (2020-02-29)
Paola Favuzza, Manuel de Lera Ruiz, Jennifer K Thompson, Tony Triglia, Anna Ngo, Ryan W J Steel, Marissa Vavrek, Janni Christensen, Julie Healer, Christopher Boyce, Zhuyan Guo, Mengwei Hu, Tanweer Khan, Nicholas Murgolo, Lianyun Zhao, Jocelyn Sietsma Penington, Kitsanapong Reaksudsan, Kate Jarman, Melanie H Dietrich, Lachlan Richardson, Kai-Yuan Guo, Sash Lopaticki, Wai-Hong Tham, Matthias Rottmann, Tony Papenfuss, Jonathan A Robbins, Justin A Boddey, Brad E Sleebs, Hélène Jousset Sabroux, John A McCauley, David B Olsen, Alan F Cowman
ABSTRACT
Artemisin combination therapy (ACT) is the main treatment option for malaria, which is caused by the intracellular parasite Plasmodium. However, increased resistance to ACT highlights the importance of finding new drugs. Recently, the aspartic proteases Plasmepsin IX and X (PMIX and PMX) were identified as promising drug targets. In this study, we describe dual inhibitors of PMIX and PMX, including WM382, that block multiple stages of the Plasmodium life cycle. We demonstrate that PMX is a master modulator of merozoite invasion and direct maturation of proteins required for invasion, parasite development, and egress. Oral administration of WM382 cured mice of P. berghei and prevented blood infection from the liver. In addition, WM382 was efficacious against P. falciparum asexual infection in humanized mice and prevented transmission to mosquitoes. Selection of resistant P. falciparum in vitro was not achievable. Together, these show that dual PMIX and PMX inhibitors are promising candidates for malaria treatment and prevention.
Sigma-Aldrich
N-Hydroxysuccinimidyl-Sepharose 4 Fast Flow, matrix fast-flow highly cross-linked 4% beaded agarose
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, 1000 units/mL
Sigma-Aldrich
N-[(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethyloxycarbonyl]-1,8-diamino-3,6-dioxaoctane, for Copper-free Click Chemistry
Sigma-Aldrich
β-Secretase human, recombinant, expressed in HEK 293 cells (C-terminal FLAG tagged), ≥10,000 units/mg protein