Merck
  • Targeting gene therapy for hepatocarcinoma cells with the E. coli purine nucleoside phosphorylase suicide gene system directed by a chimeric alpha-fetoprotein promoter.
Targeting gene therapy for hepatocarcinoma cells with the E. coli purine nucleoside phosphorylase suicide gene system directed by a chimeric alpha-fetoprotein promoter.
Cancer letters (2008-04-15)
Xiaokun Cai, Junli Zhou, Ying Chang, Xuemei Sun, Peiyuan Li, Junsheng Lin
ABSTRACT
For hepatocarcinoma (HCC) gene therapy, the tumoricidal efficacy and selective expression of therapeutic gene remain two major challenges. The Escherichia coli (E. coli) purine nucleoside phosphorylase (PNP)/9-(2-deoxy-beta-dribofuranosyl)-6-methylpurine (MeP-dR) suicide gene system exhibits excellent anti-tumor effects, indicating this system directed by a HCC-specific promoter would offer a possibility of targeting gene therapy for HCC. To test this hypothesis, here, we prepared a plasmid (p[HRE]AF/PNP) containing the E. coli PNP/MeP-dR system and a chimeric human alpha-fetoprotein (AFP) promoter, [HRE]AF. We introduced this plasmid into AFP-positive and low-AFP-generating human HCC cells, and evaluated its therapeutic effects on both human HCC cell lines. In the presence of hypoxia, the E. coli PNP gene directed by the [HRE]AF promoter were HCC-specifically expressed in two human HCC cell lines and, moreover, the [HRE]AF-PNP/MeP-dR therapy would yield significant and selective cytotoxicity in both AFP-positive and low-AFP-generating HCC cells. Our findings suggest the [HRE]AF-PNP/MeP-dR therapy has worthy potentialities as an effective strategy for targeting therapy of AFP-positive, and especially AFP-negative or low-AFP-generating HCC.
Sigma-Aldrich
6-Methylpurine, ≥99%