Merck
  • Long-Term Expansion of Pancreatic Islet Organoids from Resident Procr+ Progenitors.
Long-Term Expansion of Pancreatic Islet Organoids from Resident Procr+ Progenitors.
Cell (2020-03-24)
Daisong Wang, Jingqiang Wang, Lanyue Bai, Hong Pan, Hua Feng, Hans Clevers, Yi Arial Zeng
ABSTRACT
It has generally proven challenging to produce functional β cells in vitro. Here, we describe a previously unidentified protein C receptor positive (Procr+) cell population in adult mouse pancreas through single-cell RNA sequencing (scRNA-seq). The cells reside in islets, do not express differentiation markers, and feature epithelial-to-mesenchymal transition characteristics. By genetic lineage tracing, Procr+ islet cells undergo clonal expansion and generate all four endocrine cell types during adult homeostasis. Sorted Procr+ cells, representing ∼1% of islet cells, can robustly form islet-like organoids when cultured at clonal density. Exponential expansion can be maintained over long periods by serial passaging, while differentiation can be induced at any time point in culture. β cells dominate in differentiated islet organoids, while α, δ, and PP cells occur at lower frequencies. The organoids are glucose-responsive and insulin-secreting. Upon transplantation in diabetic mice, these organoids reverse disease. These findings demonstrate that the adult mouse pancreatic islet contains a population of Procr+ endocrine progenitors.
Sigma-Aldrich
TERGITOL solution, Type NP-40, 70% in H2O
Sigma-Aldrich
Anti-Pancreatic polypeptide/PPY antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
Monoclonal Anti-Insulin antibody produced in mouse, clone K36AC10, ascites fluid
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Monoclonal Anti-Glucagon antibody produced in mouse, clone K79bB10, ascites fluid
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Gold antibody produced in goat, affinity isolated antibody, aqueous glycerol suspension, 10 nm (colloidal gold)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Anti-SST antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Potassium hexacyanoferrate(II) trihydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, ≥180 USP units/mg