Merck
  • UBAP2L Forms Distinct Cores that Act in Nucleating Stress Granules Upstream of G3BP1.
UBAP2L Forms Distinct Cores that Act in Nucleating Stress Granules Upstream of G3BP1.
Current biology : CB (2020-01-21)
Luca Cirillo, Adeline Cieren, Sofia Barbieri, Anthony Khong, Françoise Schwager, Roy Parker, Monica Gotta
ABSTRACT
Stress granules (SGs) are membraneless organelles that form in eukaryotic cells after stress exposure [1] (reviewed in [2-4]). Following translation inhibition, polysome disassembly releases 48S preinitiation complexes (PICs). mRNA, PICs, and other proteins coalesce in SG cores [1, 5-7]. SG cores recruit a dynamic shell, whose properties are dominated by weak interactions between proteins and RNAs [8-10]. The structure and assembly of SGs and how different components contribute to their formation are not fully understood. Using super-resolution and expansion microscopy, we find that the SG component UBAP2L [11, 12] and the core protein G3BP1 [5, 11-13] occupy different domains inside SGs. UBAP2L displays typical properties of a core protein, indicating that cores of different compositions coexist inside the same granule. Consistent with a role as a core protein, UBAP2L is required for SG assembly in several stress conditions. Our reverse genetic and cell biology experiments suggest that UBAP2L forms granules independent of G3BP1 and 2 but does not interfere with stress-induced translational inhibition. We propose a model in which UBAP2L is an essential SG nucleator that acts upstream of G3BP1 and 2 and facilitates G3BP1 core formation and SG assembly and growth.
Sigma-Aldrich
Sodium acrylate, 97%
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
CRISPR/Cas9 Products and Services, Design and order CRISPR gRNA, Cas9, screening libraries, controls and companion products. Formats include plant, lentivirus, IVT-RNA, plasmid, synthetic, and protein.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
N,N′-Methylenebisacrylamide solution, suitable for electrophoresis, 2% in H2O
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-Puromycin Antibody, clone 12D10, clone 12D10, from mouse
Sigma-Aldrich
Pd-PEPPSI-IPent catalyst, ≥95%