Merck
  • Generation of Transplantable Three-Dimensional Hepatic-Patch to Improve the Functionality of Hepatic Cells In Vitro and In Vivo.
Generation of Transplantable Three-Dimensional Hepatic-Patch to Improve the Functionality of Hepatic Cells In Vitro and In Vivo.
Stem cells and development (2019-12-21)
Fatemeh Nobakht Lahrood, Mona Saheli, Zahra Farzaneh, Payam Taheri, Mahshad Dorraj, Hossein Baharvand, Massoud Vosough, Abbas Piryaei
ABSTRACT
Cell therapy and tissue engineering (TE) are considered alternative therapeutic approaches to organ transplantation. Since cell therapy approaches achieved little success for liver failure treatment, liver TE is considered a more promising alternative. In this study, we produced a liver tissue equivalent (called "liver-derived extracellular matrix scaffold [LEMS]-Patch") by co-culture of human bone marrow stromal cells, human umbilical vein endothelial cells, and a hepatoma cell line, Huh7, within an artificial three-dimensional liver-extracellular matrix scaffold. The results showed significant increase in the liver-specific gene expression and hepatic functions, in terms of albumin (ALB) and fibrinogen secretion, urea production, and cytochrome inducibility in the LEMS-Patch compared to controls. In addition, transplanted LEMS-Patch was successfully incorporated into the recipient liver of acute liver failure mice and produced human ALB. Consequently, our data demonstrated that the generated LEMS-Patch could be used as a good platform for functional improvement of hepatic cells in vitro and in vivo.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
3,3′-Methylene-bis(4-hydroxycoumarin)
Sigma-Aldrich
Alcian Blue 8GX, powder or crystals, certified by the BSC
Sigma-Aldrich
Fast Green FCF, certified by the BSC