Merck
  • Strain-promoted copper-free "click" chemistry for 18F radiolabeling of bombesin.
Strain-promoted copper-free "click" chemistry for 18F radiolabeling of bombesin.
Angewandte Chemie (International ed. in English) (2011-10-01)
Lachlan S Campbell-Verduyn, Leila Mirfeizi, Anne K Schoonen, Rudi A Dierckx, Philip H Elsinga, Ben L Feringa
Sigma-Aldrich
Dibenzocyclooctyne-PEG4-acid, for Copper-free Click Chemistry
Sigma-Aldrich
Dibenzocyclooctyne-sulfo-N-hydroxysuccinimidyl ester, for Copper-free Click Chemistry
Sigma-Aldrich
Dibenzocyclooctyne-acid, 95%, storage temp.:-20°C