Merck
  • Picolinate-appended tacn complexes for bimodal imaging: Radiolabeling, relaxivity, photophysical and electrochemical studies.
Picolinate-appended tacn complexes for bimodal imaging: Radiolabeling, relaxivity, photophysical and electrochemical studies.
Journal of inorganic biochemistry (2020-01-18)
Amaury Guillou, Margaux Galland, Amandine Roux, Balázs Váradi, Réka Anna Gogolák, Patricia Le Saëc, Alain Faivre-Chauvet, Maryline Beyler, Christophe Bucher, Gyula Tircsó, Véronique Patinec, Olivier Maury, Raphaël Tripier
ABSTRACT
Based on our previous works involving two 1,4,7-triazacyclononane (tacn)-based ligands Hno2py1pa (1-Picolinic acid-4,7-bis(pyridin-2-ylmethyl)-1,4,7-triazacyclononane) and Hno1pa (1-Picolinic acid-1,4,7-triazacyclononane), we report here the synthesis of analogues bearing picolinate-based π-conjugated ILCT (Intra-Ligand Charge Transfer) transition antenna (HL1, HL2), using regiospecific N-functionalization of the tacn skeleton and their related transition metal complexes (e.g. Cu2+, Zn2+ and Mn2+). Coordination properties as well as their photophysical and electrochemical properties were investigated in order to quantify the impact of such antenna on the luminescent or relaxometric properties of the complexes. The spectroscopic properties of the targeted ligands and metal complexes have been studied using UV-Vis absorption and fluorescence spectrocopies. While the zinc complex formed with HL1 possesses a moderate quantum yield of 5%, complexation of Cu2+ led to an extinction of the luminescence putatively attributed to a photo-induced electron transfer, as supported by spectroscopic and electrochemical evidences. The [Mn(L2)]+ complex is characterized by a fluorescence quantum yield close to 8% in CH2Cl2. The potential interest of such systems as bimodal probes has been assessed from radiolabeling experiments conducted on HL1 and 64Cu2+ as well as confocal microscopy analyses and from relaxometric studies carried out on the cationic [Mn(L2)]+ complex. These results showed that HL1 can be used for radiolabeling, with a radiochemical conversion of 40% in 15 min at 100 °C. Finally, the relaxivity values obtained for [Mn(L2)]+, r1p = 4.80 mM-1·s-1 and r2p = 8.72 mM-1·s-1, make the Mn(II) complex an ideal candidate as a probe for Magnetic Resonance Imaging.
Sigma-Aldrich
Sodium perchlorate, ACS reagent, ≥98.0%