Merck
  • Femtosecond laser-induced blastomere fusion results in embryo tetraploidy by common metaphase plate formation.
Femtosecond laser-induced blastomere fusion results in embryo tetraploidy by common metaphase plate formation.
Experimental cell research (2020-02-07)
Alina Osychenko, Alexandr Zalessky, Artyom Astafiev, Aleksander Shakhov, Andrey Kostrov, Alexandr Krivokharchenko, Viktor Nadtochenko
ABSTRACT
The cell fusion is a widespread process, which takes place in many systems in vivo and in vitro. Fusion of cells is frequently related to tetraploidy, which can be found within natural physiological conditions, e.g., placentation, and in pathophysiological conditions, such as cancer and early pregnancy failure in humans. Here we investigate the mechanism of tetraploidization with help of femtosecond laser-induced mouse blastomere fusion by the means of Hoechst staining, GFP, BODIPY dyes and fluorescent species generated intracellularly by a femtosecond laser. We establish diffusive mixing of cytosol, whereas the large components of a cytoplasm (organelles, cytoskeleton) are poorly diffusible and are not completely mixed after cell fusion and a subsequent division. We show that mechanisms which are responsible for the formation of a common metaphase plate triggered tetraploidization in fused mouse embryos and could be a significant factor in polyploidy formation in vivo. Thus, our results suggest that microtubules play a critical role in tetraploidization.
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, with 36 mg sodium pyruvate, 50 mg streptomycin sulfate, 100 mg kanamycin monosulfate, 1000 mg glucose/L and CaCl2, liquid, 0.1 μm filtered, suitable for cell culture
Sigma-Aldrich
M16 Medium, With sodium bicarbonate and lactic acid, without penicillin and streptomycin, liquid, sterile-filtered
Sigma-Aldrich
Lectin from Phaseolus vulgaris (red kidney bean), Phytohemagglutinin PHA-M, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
bisBenzimide H 33342 trihydrochloride, ≥98% (HPLC and TLC)
Sigma-Aldrich
Tyrode′s Solution, Acidic, liquid, sterile-filtered, suitable for mouse embryo cell culture
Sigma-Aldrich
M2 medium, With HEPES, without penicillin and streptomycin, liquid, sterile-filtered, suitable for mouse embryo cell culture
Sigma-Aldrich
1,3,5,7-Tetramethyl-8-phenyl-4,4-difluoroboradiazaindacene, 97%