Merck
  • Grad-seq in a Gram-positive bacterium reveals exonucleolytic sRNA activation in competence control.
Grad-seq in a Gram-positive bacterium reveals exonucleolytic sRNA activation in competence control.
The EMBO journal (2020-04-01)
Jens Hör, Geneviève Garriss, Silvia Di Giorgio, Lisa-Marie Hack, Jens T Vanselow, Konrad U Förstner, Andreas Schlosser, Birgitta Henriques-Normark, Jörg Vogel
ABSTRACT
RNA-protein interactions are the crucial basis for many steps of bacterial gene expression, including post-transcriptional control by small regulatory RNAs (sRNAs). In stark contrast to recent progress in the analysis of Gram-negative bacteria, knowledge about RNA-protein complexes in Gram-positive species remains scarce. Here, we used the Grad-seq approach to draft a comprehensive landscape of such complexes in Streptococcus pneumoniae, in total determining the sedimentation profiles of ~ 88% of the transcripts and ~ 62% of the proteins of this important human pathogen. Analysis of in-gradient distributions and subsequent tag-based protein capture identified interactions of the exoribonuclease Cbf1/YhaM with sRNAs that control bacterial competence for DNA uptake. Unexpectedly, the nucleolytic activity of Cbf1 stabilizes these sRNAs, thereby promoting their function as repressors of competence. Overall, these results provide the first RNA/protein complexome resource of a Gram-positive species and illustrate how this can be utilized to identify new molecular factors with functions in RNA-based regulation of virulence-relevant pathways.
Sigma-Aldrich
Proteomics Dynamic Range Standard Set, Protein Mass Spectrometry Calibration Standard
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)