Merck
  • Functional culture and in vitro genetic and small-molecule manipulation of adult mouse cardiomyocytes.
Functional culture and in vitro genetic and small-molecule manipulation of adult mouse cardiomyocytes.
Communications biology (2020-05-13)
Neal I Callaghan, Shin-Haw Lee, Sina Hadipour-Lakmehsari, Xavier A Lee, M Ahsan Siraj, Amine Driouchi, Christopher M Yip, Mansoor Husain, Craig A Simmons, Anthony O Gramolini
ABSTRACT
Primary adult cardiomyocyte (aCM) represent the mature form of myocytes found in the adult heart. However, culture of aCMs in particular is challenged by poor survival and loss of phenotype, rendering extended in vitro experiments unfeasible. Here, we establish murine aCM culture methods that enhance survival and maintain sarcomeric structure and Ca2+ cycling to enable physiologically relevant contractile force measurements. We also demonstrate genetic and small-molecule manipulations that probe mechanisms underlying myocyte functional performance. Together, these refinements to aCM culture present a toolbox with which to advance our understanding of myocardial physiology.
Sigma-Aldrich
Cysteamine, ~95%
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (HPLC), powder
Sigma-Aldrich
4-Nitro-3-(trifluoromethyl)phenol, 99% (GC)
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type X-S, lyophilized powder, 100,000-250,000 units/g solid (without added oxygen)
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Sigma-Aldrich
Polybrene Infection / Transfection Reagent, A highly efficient method of gene transfer into mammalian cells leveraging infection with retroviral vectors.
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Monoclonal Anti-α-Actinin (Sarcomeric) antibody produced in mouse, clone EA-53, ascites fluid
Sigma-Aldrich
Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein