- Subtype-Directed Differentiation of Human iPSCs into Atrial and Ventricular Cardiomyocytes.
Subtype-Directed Differentiation of Human iPSCs into Atrial and Ventricular Cardiomyocytes.
STAR protocols (2020-10-29)
Mandy Kleinsorge, Lukas Cyganek
PMID33111079
ABSTRACT
The generation of homogeneous populations of subtype-specific cardiomyocytes derived from human induced pluripotent stem cells (hiPSCs) is crucial in cardiovascular disease modeling as well as in drug discovery and cardiotoxicity screenings. This protocol describes a simple, robust, and efficient monolayer-based differentiation of hiPSCs into defined atrial and ventricular cardiomyocytes. For complete details on the use and execution of this protocol, please refer to Cyganek et al., 2018.
Sigma-Aldrich
Anti-RYR2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-α-Actinin (Sarcomeric) antibody produced in mouse, clone EA-53, ascites fluid
Sigma-Aldrich
Y-27632-CAS 331752-47-7-Calbiochem, Y-27632A, CAS 331752-47-7, is a cell-permeable, reversible, inhibitor of Rho kinases (Ki = 140 nM for p160ROCK). Enhances survival & cloning efficiency of ESC without affecting their pluripotency.
Sigma-Aldrich
Wnt Antagonist II, IWP-2, The Wnt Antagonist II, IWP-2, also referenced under CAS 686770-61-6, controls the biological activity of Wnt. This small molecule/inhibitor is primarily used for Cancer applications.