Merck
The DNA sequence and biological annotation of human chromosome 1.
Nature (2006-05-20)
S G Gregory, K F Barlow, K E McLay, R Kaul, D Swarbreck, A Dunham, C E Scott, K L Howe, K Woodfine, C C A Spencer, M C Jones, C Gillson, S Searle, Y Zhou, F Kokocinski, L McDonald, R Evans, K Phillips, A Atkinson, R Cooper, C Jones, R E Hall, T D Andrews, C Lloyd, R Ainscough, J P Almeida, K D Ambrose, F Anderson, R W Andrew, R I S Ashwell, K Aubin, A K Babbage, C L Bagguley, J Bailey, H Beasley, G Bethel, C P Bird, S Bray-Allen, J Y Brown, A J Brown, D Buckley, J Burton, J Bye, C Carder, J C Chapman, S Y Clark, G Clarke, C Clee, V Cobley, R E Collier, N Corby, G J Coville, J Davies, R Deadman, M Dunn, M Earthrowl, A G Ellington, H Errington, A Frankish, J Frankland, L French, P Garner, J Garnett, L Gay, M R J Ghori, R Gibson, L M Gilby, W Gillett, R J Glithero, D V Grafham, C Griffiths, S Griffiths-Jones, R Grocock, S Hammond, E S I Harrison, E Hart, E Haugen, P D Heath, S Holmes, K Holt, P J Howden, A R Hunt, S E Hunt, G Hunter, J Isherwood, R James, C Johnson, D Johnson, A Joy, M Kay, J K Kershaw, M Kibukawa, A M Kimberley, A King, A J Knights, H Lad, G Laird, S Lawlor, D A Leongamornlert, D M Lloyd, J Loveland, J Lovell, M J Lush, R Lyne, S Martin, M Mashreghi-Mohammadi, L Matthews, N S W Matthews, S McLaren, S Milne, S Mistry, M J F Moore, T Nickerson, C N O'Dell, K Oliver, A Palmeiri, S A Palmer, A Parker, D Patel, A V Pearce, A I Peck, S Pelan, K Phelps, B J Phillimore, R Plumb, J Rajan, C Raymond, G Rouse, C Saenphimmachak, H K Sehra, E Sheridan, R Shownkeen, S Sims, C D Skuce, M Smith, C Steward, S Subramanian, N Sycamore, A Tracey, A Tromans, Z Van Helmond, M Wall, J M Wallis, S White, S L Whitehead, J E Wilkinson, D L Willey, H Williams, L Wilming, P W Wray, Z Wu, A Coulson, M Vaudin, J E Sulston, R Durbin, T Hubbard, R Wooster, I Dunham, N P Carter, G McVean, M T Ross, J Harrow, M V Olson, S Beck, J Rogers, D R Bentley, R Banerjee, S P Bryant, D C Burford, W D H Burrill, S M Clegg, P Dhami, O Dovey, L M Faulkner, S M Gribble, C F Langford, R D Pandian, K M Porter, E Prigmore
ABSTRACT
The reference sequence for each human chromosome provides the framework for understanding genome function, variation and evolution. Here we report the finished sequence and biological annotation of human chromosome 1. Chromosome 1 is gene-dense, with 3,141 genes and 991 pseudogenes, and many coding sequences overlap. Rearrangements and mutations of chromosome 1 are prevalent in cancer and many other diseases. Patterns of sequence variation reveal signals of recent selection in specific genes that may contribute to human fitness, and also in regions where no function is evident. Fine-scale recombination occurs in hotspots of varying intensity along the sequence, and is enriched near genes. These and other studies of human biology and disease encoded within chromosome 1 are made possible with the highly accurate annotated sequence, as part of the completed set of chromosome sequences that comprise the reference human genome.
Sigma-Aldrich
Phospholipase A2 from porcine pancreas, ammonium sulfate suspension, ≥600 units/mg protein
Sigma-Aldrich
Phospholipase A2 from bovine pancreas, lyophilized powder, ≥20 units/mg protein
Sigma-Aldrich
Neuron-specific enolase from human brain, ≥95% (SDS-PAGE), buffered aqueous solution
Sigma-Aldrich
6-Phosphogluconic Dehydrogenase from yeast, lyophilized powder, ≥3.0 units/mg solid
Sigma-Aldrich
Galactosyltransferase from bovine milk, lyophilized powder
Sigma-Aldrich
Phosphatase, Acid from wheat germ, ≥0.4 unit/mg solid
Sigma-Aldrich
MNK1 active human, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE)
Sigma-Aldrich
Cyclooxygenase 1 from sheep, glycerol solution, ≥1500 units/mg protein
Sigma-Aldrich
Myokinase from rabbit muscle, ammonium sulfate suspension, 1,500-3,000 units/mg protein (biuret)
Sigma-Aldrich
Nucleoside 5′-Diphosphate Kinase from bovine liver, buffered aqueous glycerol solution, ≥1,000 units/mg protein (biuret)
Sigma-Aldrich
Phosphatase, Acid from sweet potato, ammonium sulfate suspension, ≥10.0 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Phosphoglucomutase from rabbit muscle, ammonium sulfate suspension, ≥100 units/mg protein
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥0.5 unit/mg solid
Sigma-Aldrich
Enolase from baker′s yeast (S. cerevisiae), lyophilized powder, ≥50 units/mg protein
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥3.0 units/mg solid
Sigma-Aldrich
Phospholipase A2 from honey bee venom (Apis mellifera), salt-free, lyophilized powder, 600-2400 units/mg protein
Sigma-Aldrich
L-Glutamine Synthetase from Escherichia coli, lyophilized powder, 400-2,000 units/mg protein
Sigma-Aldrich
5′-Nucleotidase human, recombinant, expressed in CHO cells, vial of 6-12 μg