Merck
  • Paramagnetic Cobalt(II) Complexes with Cyclam Derivatives: Toward 19F MRI Contrast Agents.
Paramagnetic Cobalt(II) Complexes with Cyclam Derivatives: Toward 19F MRI Contrast Agents.
Inorganic chemistry (2020-07-08)
Jan Blahut, Ladislav Benda, Jan Kotek, Guido Pintacuda, Petr Hermann
ABSTRACT
In order to develop novel, more efficient, and/or selective contrast agents for magnetic resonance imaging (MRI), different modi operandi are explored as alternatives to water-relaxation enhancement. In this work, cobalt(II/III) complexes of bis(N-trifluoroethyl)cyclam derivatives with two acetate or two phosphonate pendant arms, H2te2f2a and H4te2f2p, were prepared and investigated. X-ray diffraction structures confirmed octahedral coordination with a very stable trans-III cyclam conformation and with fluorine atoms located about 5.3 Å from the metal center. The Co(II) complexes are kinetically inert, decomposing slowly even in 1 M aqueous HCl at 80 °C. The Co(II) complexes exhibited well-resolved paramagnetically shifted NMR spectra. These were interpreted with the help of quantum chemistry calculations. The 13C NMR shifts of the trans-[CoII(te2f2p)]2- complex were successfully assigned based on spin density delocalization within the ligand molecule. The obtained spin density also helps to describe d-metal-induced NMR relaxation properties of 19F nuclei, including the contribution of a Fermi contact relaxation mechanism. The paramagnetic complexes show convenient relaxation properties to be used as 19F MRI contrast agents.
Sigma-Aldrich
1,4,8,11-Tetraazacyclotetradecane, 98% (GC)