Merck
Persistent Roseoloviruses Infection in Adult Patients with Epilepsy.
Brain sciences (2020-05-15)
Santa Rasa-Dzelzkaleja, Sabine Gravelsina, Svetlana Chapenko, Zaiga-Nora Krukle, Simons Svirskis, Normunds Suna, Elena Kashuba, Guntis Karelis, Modra Murovska
ABSTRACT
Human herpesviruses (HHV)-6A, HHV-6B and HHV-7 are considered to be involved in the pathogenesis of epilepsy, a common neurological disorder. The objective of this study was to determine the association of roseoloviruses infection with epilepsy. 53 epilepsy patients and 104 ordinary blood donors were analyzed to determine presence of virus-specific antibodies by enzyme-linked immunosorbent assay (ELISA) and immunofluorescence assay (IFA), genomic sequences, viral load and gene expression by polymerase chain reactions (PCRs) and restriction analysis, HHV-6 protein expression by IFA and level of cytokines by ELISA. Roseoloviruses genomic sequences in DNA samples from whole blood were found in 86.8% of patients versus 54.8% of controls and active infection was revealed only in patients with epilepsy (19.6% of roseolovirus-positive patients). Significantly higher viral load and more frequent gene expression was detected in patients compared to the controls. HHV-6-encoded protein expression was demonstrated in 53.3% of patients with previously detected HHV-6 DNA. Changes in level of cytokines were determined in patients with elevated viral load compared to the patients without elevated viral loads and to the controls. Results on frequent active HHV-6 and HHV-7 infection in epilepsy patient' peripheral blood indicate on possible involvement of these viruses in the disease development.
Sigma-Aldrich
Human IgG ELISA Kit, for serum, plasma
Pricing and availability is not currently available.