Merck
  • Estrogen-ERα signaling and DNA hypomethylation co-regulate expression of stem cell protein PIWIL1 in ERα-positive endometrial cancer cells.
Estrogen-ERα signaling and DNA hypomethylation co-regulate expression of stem cell protein PIWIL1 in ERα-positive endometrial cancer cells.
Cell communication and signaling : CCS (2020-06-07)
Zheng Chen, Hua-Jing Yang, Qin Lin, Min-Jiao Zhu, Ying-Ying Yu, Xiao-Ying He, Xiao-Ping Wan
ABSTRACT
We previously identified PIWIL1 as an oncogene involved in endometrial carcinogenesis. However, the mechanism of Piwil1 mediated regulation of tumorigenesis remains poorly understood. The expression levels of target genes in endometrial cancer cells were detected by quantitative reverse transcription-PCR (RT-qPCR) and western blotting. Up- or down-regulation of ERα or PIWIL1 was achieved by transient transfection with expressing plasmids or short hairpin RNA (shRNA). Dual-luciferase reporter assays and chromatin immunoprecipitation (ChIP) were used to demonstrate the ERα bound to the half estrogen response element (half-ERE) located in PIWIL1 promoter. The expression of PIWIL1 and ERα in endometrial carcinoma tissues were investigated using immunohistochemistry and RT-qPCR. The proliferation ability of cancer cells were evaluated by MTT. Methylation status of the PIWIL1 promoter was detected by bisulfite sequencing PCR (BSP). In the present study, we found that PIWIL1 mediated E2-stimulated cancer cell proliferation. In ERα-positive endometrial cancer cells, we demonstrated that estrogen-ERα signaling significantly up-regulated the expression of PIWIL1, which was mediated by binding of the ERα onto the PIWIL1 promoter. Furthermore, we found that a half-ERE in the PIWIL1 promoter was essential for ERα binding. The PIWIL1 promoter was hypomethylated in ERα-positive endometrial cancer cells. Treatment with 5-aza-deoxycytidine (5-aza-dC) could up-regulate PIWIL1 expression. These findings uncover a novel molecular mechanism by which estrogen-ERα signaling and DNA hypomethylation co-regulate PIWIL1 expression. These findings provide novel insights into the hormonal regulation of PIWIL1 in endometrial cancer and the PIWIL1's role in estrogen-stimulated endometrial carcinogenesis. Video Abstract. (MP4 41319 kb).
Sigma-Aldrich
MISSION® esiRNA, targeting human PIWIL1