Merck
Mechanism of Manganese Dysregulation of Dopamine Neuronal Activity.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2020-06-25)
Min Lin, Luis M Colon-Perez, Danielle O Sambo, Douglas R Miller, Joseph J Lebowitz, Felix Jimenez-Rondan, Robert J Cousins, Nicole Horenstein, Tolunay Beker Aydemir, Marcelo Febo, Habibeh Khoshbouei
ABSTRACT
Manganese exposure produces Parkinson's-like neurologic symptoms, suggesting a selective dysregulation of dopamine transmission. It is unknown, however, how manganese accumulates in dopaminergic brain regions or how it regulates the activity of dopamine neurons. Our in vivo studies in male C57BLJ mice suggest that manganese accumulates in dopamine neurons of the VTA and substantia nigra via nifedipine-sensitive Ca2+ channels. Manganese produces a Ca2+ channel-mediated current, which increases neurotransmitter release and rhythmic firing activity of dopamine neurons. These increases are prevented by blockade of Ca2+ channels and depend on downstream recruitment of Ca2+-activated potassium channels to the plasma membrane. These findings demonstrate the mechanism of manganese-induced dysfunction of dopamine neurons, and reveal a potential therapeutic target to attenuate manganese-induced impairment of dopamine transmission.SIGNIFICANCE STATEMENT Manganese is a trace element critical to many physiological processes. Overexposure to manganese is an environmental risk factor for neurologic disorders, such as a Parkinson's disease-like syndrome known as manganism. We found that manganese concentration-dependently increased the excitability of dopamine neurons, decreased the amplitude of action potentials, and narrowed action potential width. Blockade of Ca2+ channels prevented these effects as well as manganese accumulation in the mouse midbrain in vivo Our data provide a potential mechanism for manganese regulation of dopaminergic neurons.
Product Description
SAFC
L-Glutamine Solution 200 mM, 29.23 mg/mL in saline, solution, suitable for cell culture
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Sigma-Aldrich
Cytosine β-D-arabinofuranoside hydrochloride, crystalline
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Anti-Cav1.2 calcium channel Antibody, clone L57/46, clone L57/46, from mouse
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Cadmium chloride, 99.99% trace metals basis
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
BAPTA-AM, ≥95% (HPLC)
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder