Merck
  • Mediation of Cartilage Matrix Degeneration and Fibrillation by Decorin in Post-traumatic Osteoarthritis.
Mediation of Cartilage Matrix Degeneration and Fibrillation by Decorin in Post-traumatic Osteoarthritis.
Arthritis & rheumatology (Hoboken, N.J.) (2020-03-13)
Qing Li, Biao Han, Chao Wang, Wei Tong, Yulong Wei, Wei-Ju Tseng, Li-Hsin Han, X Sherry Liu, Motomi Enomoto-Iwamoto, Robert L Mauck, Ling Qin, Renato V Iozzo, David E Birk, Lin Han
ABSTRACT
To elucidate the role of decorin, a small leucine-rich proteoglycan, in the degradation of cartilage matrix during the progression of post-traumatic osteoarthritis (OA). Three-month-old decorin-null (Dcn-/- ) and inducible decorin-knockout (DcniKO ) mice were subjected to surgical destabilization of the medial meniscus (DMM) to induce post-traumatic OA. The OA phenotype that resulted was evaluated by assessing joint morphology and sulfated glycosaminoglycan (sGAG) staining via histological analysis (n = 6 mice per group), surface collagen fibril nanostructure via scanning electron microscopy (n = 4 mice per group), tissue modulus via atomic force microscopy-nanoindentation (n = 5 or more mice per group) and subchondral bone structure via micro-computed tomography (n = 5 mice per group). Femoral head cartilage explants from wild-type and Dcn-/- mice were stimulated with the inflammatory cytokine interleukin-1β (IL-1β) in vitro (n = 6 mice per group). The resulting chondrocyte response to IL-1β and release of sGAGs were quantified. In both Dcn-/- and DcniKO mice, the absence of decorin resulted in accelerated sGAG loss and formation of highly aligned collagen fibrils on the cartilage surface relative to the control (P < 0.05). Also, Dcn-/- mice developed more salient osteophytes, illustrating more severe OA. In cartilage explants treated with IL-1β, loss of decorin did not alter the expression of either anabolic or catabolic genes. However, a greater proportion of sGAGs was released to the media from Dcn-/- mouse explants, in both live and devitalized conditions (P < 0.05). In post-traumatic OA, decorin delays the loss of fragmented aggrecan and fibrillation of cartilage surface, and thus, plays a protective role in ameliorating cartilage degeneration.
Sigma-Aldrich
L-Ascorbic acid 2-phosphate sesquimagnesium salt hydrate, ≥95%
Sigma-Aldrich
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride, ≥97.0% (AT)
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, Type II-S, 1,000-2,000 units/mg dry solid
Sigma-Aldrich
Sesame oil, Antioxidant, delivery vehicle for fat-soluble compounds
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid