Merck
  • Interleukin-35 inhibits human umbilical vein endothelial cell injury induced by sera from pre-eclampsia patients by up-regulating S100A8 protein expression.
Interleukin-35 inhibits human umbilical vein endothelial cell injury induced by sera from pre-eclampsia patients by up-regulating S100A8 protein expression.
Hypertension in pregnancy (2020-03-24)
Ming Li, Lei Qian, Jianxiu Yu, Yingfen Zou
ABSTRACT
Objectives: The protective effects of interleukin(IL)-35 against injury to human umbilical vein endothelial cells (HUVECs) induced by the serum of pre-eclampsia patients were analyzed.Methods: This cross-sectional study included 24 patients with pre-eclampsia (PE) and 24 normotensive pregnant women.Results: Compared to normotensive pregnant women, patients with pre-eclampsia had lower IL-35 levels (P < 0.05). In addition, our in vitro experiments, IL-35 inhibited the PE serum-induced apoptosis of HUVECs and the levels of reactive oxygen species in HUVECs.Conclusion: Decreases in the serum IL-35 level may play an important role in the pathogenesis of endothelial dysfunction in patients with pre-eclampsia.
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human S100A8
Sigma-Aldrich
JC-1, powder or solid (Crystals)