Merck
  • Self-Healing and Mechanical Properties of Thermoplastic Polyurethane/Eugenol-Based Phenoxy Resin Blends via Exchange Reactions.
Self-Healing and Mechanical Properties of Thermoplastic Polyurethane/Eugenol-Based Phenoxy Resin Blends via Exchange Reactions.
Polymers (2020-05-02)
Jing-Yu Liang, Se-Ra Shin, Soo-Hyoung Lee, Dai-Soo Lee
ABSTRACT
The possibility of exchange reactions and thermal self-healing in blends of thermoplastic polyurethane (TPU) and phenoxy resin was investigated herein. The analyses were based on characterization obtained via differential scanning calorimetry (DSC), Fourier transform infrared spectroscopy (FTIR), dynamic mechanical analysis (DMA), and tensile test. A new phenoxy resin was synthesized from eugenol, and blends with different types of TPU were prepared to investigate the exchange reaction, thermal self-healing, and mechanical properties. The influence of phenoxy resin content on the mechanical behavior and healing efficiency was studied. Improvement of storage modulus owing to the increase of phenoxy resin content was observed. Results suggest that the exchange reaction between phenoxy- and ester-type TPU occurred during thermal treatment. However, little exchange occurred between phenoxy resin and ether-type TPU. Specifically, only ester-type TPU exhibited a significant exchange reaction in the phenoxy resin blend. Furthermore, in the presence of a catalyst (e.g., zinc acetate), the exchange reaction readily occurred, and the healing efficiency improved by the addition of the catalyst and increase in the phenoxy content.
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Sigma-Aldrich
3-Chloroperbenzoic acid, ≤77%