Merck
  • Ganoderic acid A/DM-induced NDRG2 over-expression suppresses high-grade meningioma growth.
Ganoderic acid A/DM-induced NDRG2 over-expression suppresses high-grade meningioma growth.
Clinical & translational oncology : official publication of the Federation of Spanish Oncology Societies and of the National Cancer Institute of Mexico (2019-11-17)
A Das, M Alshareef, F Henderson, J L Martinez Santos, W A Vandergrift, S M Lindhorst, A K Varma, L Infinger, S J Patel, D Cachia
ABSTRACT
N-myc downstream-regulated gene 2 (NDRG2) is down-regulated in grade-III meningioma [anaplastic meningioma (AM)] and associated with clinically aggressive behavior. Current therapies in the treatment of high-grade meningioma are lacking with limited success. This study aims to validate the effect of NDRG2-targeted therapy using structurally related bioactive triterpene compounds derived from the edible mushroom Ganoderma lucidum (ganoderic acid A:GA-A/ganoderic acid DM:GA-DM) in human AM in relevant pre-clinical models. Tissue samples from the AM tumor regions of three human patients and control non-tumor samples were used to analyze the expression pattern of NDRG2. In vitro cell culture and in vivo cell-line-derived orthotopic xenograft animal models of AM were utilized to assess efficacy of treatment with GA-A/DM. Downregulation of NDRG2 expression was observed in surgically resected high-grade meningiomas compared to normal brain. These results prompt us to use NDRG2-targeting agents GA-A/DM. In vitro results showed that 72-h treatments of 25 µM GA-A/DM induced AM cell death, upregulate NDRG2 protein expression, downregulate NDRG2 promoter methylation in meningioma cells as compared to azacitidine and decitabine, the most commonly used demethylating agents. Our results also demonstrated that GA-A/DM does not have any detrimental effect on normal human neurons and arachnoid cells. GA-A/DM promoted apoptotic factors (Bax) while suppressing MMP-9, p-P13K, p-AKT, p-mTOR, and Wnt-2 protein expression. RNAi-mediated knockdown of NDRG2 protein expression increased tumor proliferation, while forced expression of wt-NDRG2 decreased proliferation in an in vitro model. Magnetic resonance (MR) imaging and Hematoxylin (H&E) staining demonstrated gross reduction of tumor volume in GA-A/DM treated mice at 5 weeks when compared with saline-treated orthotopic AM xenografted controls. There was an overall decrease in tumor cell proliferation with increased survival in GA-A/DM-treated animals. Enzyme assays showed that GA-A/DM did not negatively impact hepatic function. GA-A/DM may be a promising natural therapeutic reagent in the treatment of AM by suppressing growth via NDRG2 modulation and altering of intracellular signal pathways. We have shown it could potentially be an effective treatment for AM with decreased cellular proliferation in vitro, decreased tumor volume and increased survival in vivo.
Sigma-Aldrich
MISSION® esiRNA, targeting human NDRG2